VisiLED LED Illumination Systems
Lieferant: SCHOTT AG Lighting and imaging
400000 400080
630-3500EA 1160 CHF
630-3500 630-3181
VisiLED LED Illumination Systems
Beleuchtung Kaltlichtquellen
Professional illumination for stereo microscopy. The SCHOTT VisiLED series is a range of advanced modular LED illumination products specially designed for stereo microscopy. Active control of the LED temperature, the option to select different controllers, and a large number of accessories enable bespoke lighting systems for specific applications.
  • Flexible options for strong contrast
  • VisiLED product line offers powerful light heads for tasks up to the highest magnification
  • Darkfield options and control of individual segments enable a number of contrasting