Foot pedal for Dino-Lite microscopes, SW-F1
Lieferant: Dino-Lite / IDCP B.V.
SW-F1
630-1821EA 99 CHF
630-1821
Foot pedal for Dino-Lite microscopes, SW-F1
Mikroskope
The SW-F1 allows users to take a picture with a simple step from their foot using the Dino-Lite or Dino-Eye digital microscopy solutions. The foot pedal is only compatible with DinoCapture 2.0 for the PC and DinoXcope for Macintosh computers. No additional software is required, simply plug in the device into your USB port and it is ready to use.
Compatible with all USB models.