Buffer, triethylammonium acetate, 1 M solution, pH 7,0, Calbiochem®, Millipore®
Lieferant: Merck Millipore Calbiochem
625718-1
625718-1EA 820 CHF
625718-1
Buffer, triethylammonium acetate, 1 M solution, pH 7,0, Calbiochem®, Millipore®
Puffer Biologische Puffer
This buffer solution is used for the purification of chemically synthesised oligonucleotides by HPLC.