VWR®, Dispenser-Spitzen, universal
Lieferant: VWR Collection
613-1032EA 0 CHF
613-1032 613-1033 613-5867 613-5868 613-5869 613-5870 613-5871 613-5872 613-5873 613-5874 613-5875 613-5877 613-5878 613-5879 613-5880 613-5881 613-5882 613-5883 613-5884
VWR®, Dispenser-Spitzen, universal
Pipettenspitzen
By using the direct displacement method, these tips work well for infectious and volatile liquids, as the fluid never will get in touch with the instrument which avoids any contamination.
  • Compatible with: VWR® Stepper and Stepper Pro, Ritter Ripette®, and Ripette® pro, Brand HandyStep®, HandyStep® electronic and HandyStep® touch*, Eppendorf® Multipette® 4780 and Multipette® plus, Gilson Repetman®, EasyStep, Minilab 100/101, STEPMATE Handrop (* without activated display/tip recognition)
  • Sustainable design based on virgin resins
  • Nine different tip sizes
More Product Information