VWR®, Pipette Tips, Pak Rak™ Express Reload System
Lieferant: VWR Collection
613-0249EA 57.3 CHF
613-0249 613-0258 613-0363 613-0631
VWR®, Pipette Tips, Pak Rak™ Express Reload System
Pipettenspitzen
These tips are made of high quality 100% pure virgin medical grade PP. Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard.
  • A wider seal ensures that tips will seal easily and consistently without leaking
  • Guaranteed free of detectable DNA, DNase/RNase, endotoxins and heavy metals
  • Graduated tips only: Graduations allow a visual check of pipetting accuracy
  • Sterile products are tested for endotoxin (pyrogen) contamination and are certified to USP standards
  • Resins are pre-tested for metal contamination