Analysewaagen, Entris® II advanced
Lieferant: Sartorius Balances
BCA124-1S BCA124I-1S BCA224-1S BCA224I-1S BCA324I-1S BCA64-1S BCA64I-1S BCA124I-1CEU BCA224I-1CEU BCA324I-1CEU BCA64I-1CEU
611-4844EA 4470 CHF
611-4844 611-4845 611-4852 611-4853 611-4858 611-4869 611-4870 611-4883 611-4891 611-4897 611-4909
Analysewaagen, Entris® II advanced
Waagen
Entris® II Advanced analytical balances feature isoCAL, intuitive graphic touch display and 13 built-in applications. Highly accurate results are guaranteed via the monolithic weigh cell technology. High chemical resistance is ensured by using parts made from hard-wearing PBT, stainless steel and glass. Integrated protection systems increase reliability of weighing results: Three configurable levels determine valid weighing data and ensure only valid data is transferred to external devices.
  • Fast stabilisation time thanks to state-of-the-art weighing
More Product Information
Order Now
TECHNISCHE DATEN
Environmentally Preferable