Kieselgel, Porengröße: 100 Å 35-70 mesh, high_purity Chromatography_column, Supelco®

Lieferant: Merck


MQ100

60746-1KG
60746-1KGEA 167 CHF
60746-1KG
Kieselgel, Porengröße: 100 Å 35-70 mesh, high_purity Chromatography_column, Supelco®
Silica gel
Formel: SiO₂
MW: 60.08 g/mol
Siedepunkt: 2230
Melting Pt: 1710
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011232
CAS-Nummer: 112926-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=333d0ad5223a90e106e344c5eb00aed228a4b420&vid=7e7df3f1f3902790d7de4a09d3dc485c6b8db31f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR