Kieselgel, Porengröße: 60 Å 230-400 mesh, high_purity Chromatography_flash, Supelco®

Lieferant: Merck


MQ100

60737-2.5KG 60737-500G 60737-1KG 60737-25KG 60737-5KG
60737-2.5KGEA 640 CHF
60737-2.5KG 60737-500G 60737-1KG 60737-25KG 60737-5KG
Kieselgel, Porengröße: 60 Å 230-400 mesh, high_purity Chromatography_flash, Supelco®
Silica gel
Formel: SiO₂
MW: 60.08 g/mol
Siedepunkt: 2230
Melting Pt: 1710
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011232
CAS-Nummer: 112926-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=672d590b438a0f58c5a72b4e431a2384f386ee6f&vid=19a4617ff7fa36beb691c559314943d13c7d04e5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR