Mouse IgG2b Isotype Control [clone: MPC-11] (PE (Phycoerythrin)-Cy7®)
Lieferant: Tonbo Biosciences
60-4732-U100
60-4732-U100EA 198 CHF
60-4732-U100
Mouse IgG2b Isotype Control [clone: MPC-11] (PE (Phycoerythrin)-Cy7®)
Antikörper
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.