2-Propanol, Finyte® Microelectronic, J.T.Baker®

Lieferant: Avantor

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

M.O.C. Category R

5873-15EA 186 CHF
5873-15
2-Propanol, Finyte® Microelectronic, J.T.Baker®
2-Propanol

In the Avantor Electronic Materials range, we have solvents, acids and bases in CMOS and Finyte/VLSI quality, as well as Performance Materials like photoresist- and residue-removers, selective etchants, BEOL and FEOL.
Avantor Performance Materials portfolio of production-proven chemistries, customizable chemistry platforms and world-class applications centers are designed to help you meet your process improvement goals faster and speed up your ramp time to the next node.

Formel: (CH₃)₂CHOH
MW: 60.1 g/mol
Siedepunkt: 82
Melting Pt: -89
Flash-Pt: 12
MDL Number: MFCD00011674
CAS-Nummer: 67-63-0
UN: 1219
ADR: 3 II
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

For Microelectronic Use = 99.9 %
Assay (CH₃CHOHCH₃) ≥ 99.9 %
Color (APHA) ≤ 6
Residue after Evaporation ≤ 3000.0 ppb
Solubility in H₂O Passes Test
Water (H₂O)(by Karl Fischer titrn) ≤ 0.05 %
Acidity (µeq/g) ≤ 0.2
Alkalinity (µeq/g) ≤ 0.1
Chloride (Cl) ≤ 0.2 ppm
Phosphate (PO₄) ≤ 75.0 ppb
Trace Impurities - Aluminum (Al) ≤ 10.0 ppb
Arsenic and Antimony (as As) ≤ 10.0 ppb
Trace Impurities - Barium (Ba) ≤ 10.0 ppb
Trace Impurities - Beryllium (Be) ≤ 10.0 ppb
Trace Impurities - Bismuth (Bi) ≤ 10.0 ppb
Trace Impurities - Boron (B) ≤ 3.0 ppb
Trace Impurities - Cadmium (Cd) ≤ 10.0 ppb
Trace Impurities - Calcium (Ca) ≤ 10.0 ppb
Trace Impurities - Chromium (Cr) ≤ 10.0 ppb
Trace Impurities - Cobalt (Co) ≤ 10.0 ppb
Trace Impurities - Copper (Cu) ≤ 10.0 ppb
Trace Impurities - Gallium (Ga) ≤ 10.0 ppb
Trace Impurities - Germanium (Ge) ≤ 10.0 ppb
Trace Impurities - Gold (Au) ≤ 5.0 ppb
Trace Impurities - Iron (Fe) ≤ 5.0 ppb
Trace Impurities - Lead (Pb) ≤ 10.0 ppb
Trace Impurities - Lithium (Li) ≤ 10.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 10.0 ppb
Trace Impurities - Manganese (Mn) ≤ 10.0 ppb
Trace Impurities - Molybdenum (Mo) ≤ 10.0 ppb
Trace Impurities - Nickel (Ni) ≤ 10.0 ppb
Trace Impurities - Niobium (Nb) ≤ 10.0 ppb
Trace Impurities - Potassium (K) ≤ 10.0 ppb
Trace Impurities - Silicon (Si) ≤ 10.0 ppb
Trace Impurities - Silver (Ag) ≤ 5.0 ppb
Trace Impurities - Sodium (Na) ≤ 10.0 ppb
Trace Impurities - Strontium (Sr) ≤ 10.0 ppb
Trace Impurities - Tantalum (Ta) ≤ 10.0 ppb
Trace Impurities - Thallium (Tl) ≤ 10.0 ppb
Trace Impurities - Tin (Sn) ≤ 10.0 ppb
Trace Impurities - Titanium (Ti) ≤ 10.0 ppb
Trace Impurities - Vanadium (V) ≤ 10.0 ppb
Trace Impurities - Zinc (Zn) ≤ 10.0 ppb
Trace Impurities - Zirconium (Zr) ≤ 10.0 ppb
Particle Count - 0.5 µm and greater ≤ 25 par/ml
Particle Count - 1.0 µm and greater ≤ 8 par/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR