Dithiothreitol (DTT, Cleland's Reagenz), SPUTOLYSIN®

Lieferant: Merck Millipore Calbiochem

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

560000-1 560000-10
560000-1EA 1020 CHF
560000-1 560000-10
Dithiothreitol (DTT, Cleland's Reagenz), SPUTOLYSIN®
Dithiothreitol (DTT, Cleland's Reagenz)

Spotolysin reagent is a concentrated solution of Dithiothreitol in phosphate buffer. Mainly for the isolation of epithelial cells, pathogenic or saprophytic bacteria, fungi, and yeasts from sputum.


  • Epethelial cell isolation
  • Concentrate of dithiothreitol (DTT) in phosphate buffer
  • Isolation from sputum
  • pH 6.5 - 7.5


Each vial is sufficient to make 100 ml of working solution.

Formel: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004877
CAS-Nummer: 3483-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR