Capillary columns, TraceGOLD™ TG-35MS
Lieferant: Thermo Fisher Scientific
26094-2130 26094-2230 26094-1430 26094-1420 26094-1300 26094-2980 26094-3360 26094-2240
555-8754EA 753 CHF
555-8754 555-8753 555-8757 555-8758 555-8760 555-8745 555-8743 555-8752
Capillary columns, TraceGOLD™ TG-35MS
Chromatographiesäulen
These GC columns have a higher phenyl content for useful elution order and retention time changes.
  • Mid-polarity phase, 35% diphenyl/65% dimethyl polysiloxane
  • Equivalent to USP G42 phase
Ideal for organochlorine pesticides and herbicides, pharmaceuticals, sterols, phthalate esters etc.