HPLC columns, GlycanPac™ AXR-1
Lieferant: Thermo Fisher Scientific
088135 088136
555-7334EA 2290 CHF
555-7334 555-7335
HPLC columns, GlycanPac™ AXR-1
Chromatographiesäulen
The GlycanPac™ AXR-1 column uses volatile aqueous buffers (e.g., ammonium acetate or ammonium formate) and acetonitrile, presenting the eluting glycans ready for introduction into MS instruments. Designed and tested for HPLC and UHPLC methods using either fluorescence or MS detection applications. Ultra-high resolution and selectivity of glycans are attributed to the GlycanPac™ AXR-1 column's innovative mixed-mode surface chemistry, which combines:
  • WAX functionality for retention of negatively charged glycans, with separation in order of increasing charge
More Product Information