BioHPLC columns, DNAPac™ PA200

Lieferant: Thermo Fisher Scientific
063419 063421 063423 063425 088780 088781 062998 063000
555-7283EA 1140 CHF
555-7283 555-7284 555-7285 555-7286 555-0253 555-7456 555-7323 555-7324
BioHPLC columns, DNAPac™ PA200
Chromatographiesäulen
The PA200 columns are strong anion exchange column developed to provide unsurpassed high resolution analysis and purification of synthetic oligonucleotides.

  • With secondary structures it resolve oligonucleotides
  • Assay phosphorothioate purity
  • Selectivity control with eluent pH, salt and solvent
  • pH range: 2 to 12,5
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR