Anti-c-Raf (pTyr 340 , Tyr 341 ) Rabbit Polyclonal Antibody

Lieferant: Merck Millipore (Oncogene)
553009-10
553009-10EA 1210 CHF
553009-10
Anti-c-Raf (pTyr 340 , Tyr 341 ) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: c-Raf (pTyr 340 , Tyr 341 )
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR