Glass screw top vials, 2 ml, level 2 high-throughput applications, SureSTART™
Lieferant: Thermo Fisher Scientific
6ASV9-1P 6ASV9-1PF 6ASV9-1PT 6ASV9-2P 6ASV9-2PF 6ASV9-2PT 6ASV9-S2P
548-2742EA 14.5 CHF
548-2742 548-2743 548-2744 548-2745 548-2746 548-2747 548-2748
Glass screw top vials, 2 ml, level 2 high-throughput applications, SureSTART™
Vials
Choose Thermo Scientific™ SureSTART™ 2 ml glass screw top vials, performance level 2, to ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems. Use level 2 vials for routine QA/QC applications, where the analyte levels are achievable or close to the method detection level. Available in clear and amber glass, these 2 ml screw top vials have very low metal content, extractables, and leachables and conform to high standards so that you can to meet your regulatory requirements. They areMore Product Information