Screw caps, 9 mm, level 2 high-throughput applications, SureSTART™
Lieferant: Thermo Fisher Scientific
6ASC9RT1 6ASC9RT1F 6ASC9RT1R 6ASC9RT1RF 6ASC9RT1RT 6ASC9RT1T 6ASC9ST1 6ASC9ST1F 6ASC9ST1R 6ASC9ST1RF 6ASC9ST1RT 6ASC9ST1T 6ASC9STB1 6ASC9STB1F 6ASC9STB1T 6ASC9STBS1 6ASC9STBS1F 6ASC9STBS1T
548-2724EA 34.7 CHF
548-2724 548-2725 548-2726 548-2727 548-2728 548-2729 548-2730 548-2731 548-2732 548-2733 548-2734 548-2735 548-2736 548-2737 548-2738 548-2739 548-2740 548-2741
Screw caps, 9 mm, level 2 high-throughput applications, SureSTART™
Verschlüsse Chromatographieampullen-Verschlüsse
Use Thermo Scientific™ SureSTART™ 9 mm screw caps with screw vials that have a 9 mm opening, including our SureSTART 2 ml glass screw vials and 1,7 ml glass high recovery screw microvials for <2 ml samples (level 2). Select level 2 products for high-throughput applications, when robustness and reproducibility are key.
Choose from polypropylene caps with all-purpose silicone/PTFE septa of various hardness values designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. Our AVCS™ caps incorporate advanced vial closure system technology to ensureMore Product Information