Ammoniaklösung 28 - 30% VLSI für die Elektronikindustrie, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

5358-03EA 209 CHF
5358-03
Ammoniaklösung 28 - 30% VLSI für die Elektronikindustrie, J.T.Baker®
Ammoniaklösung
Formel: NH₃
MW: 35.05 g/mol
Siedepunkt: 32…37.7 °C (1013 hPa)
Melting Pt: –60…–57.5 °C
Dichte: 0.88…0.91 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00066650
CAS-Nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Order Now

Spezifikation Testergebnisse

For Microelectronic Use Passes Test
Appearance Passes Test
Assay (as NH₃) 28.0 - 30.0 %
Color (APHA) ≤ 10
Insoluble Matter ≤ 1 ppm
Residue after Ignition ≤ 3 ppm
Carbon Dioxide (CO₂) ≤ 10 ppm
Pyridine Passes Test
Substances Reducing Permanganate Passes Test
Chloride (Cl) ≤ 0.3 ppm
Phosphate (PO₄) ≤ 0.20 ppm
Total Sulfur (as SO₄) ≤ 0.8 ppm
Trace Impurities - Aluminum (Al) ≤ 10.0 ppb
Arsenic and Antimony (as As) ≤ 30.0 ppb
Trace Impurities - Barium (Ba) ≤ 10.0 ppb
Trace Impurities - Beryllium (Be) ≤ 10.0 ppb
Trace Impurities - Bismuth (Bi) ≤ 20.0 ppb
Trace Impurities - Boron (B) ≤ 10.0 ppb
Trace Impurities - Cadmium (Cd) ≤ 5.0 ppb
Trace Impurities - Calcium (Ca) ≤ 100.0 ppb
Trace Impurities - Chromium (Cr) ≤ 5.0 ppb
Trace Impurities - Cobalt (Co) ≤ 1.0 ppb
Trace Impurities - Copper (Cu) ≤ 10.0 ppb
Trace Impurities - Gallium (Ga) ≤ 50 ppb
Trace Impurities - Germanium (Ge) ≤ 50.0 ppb
Trace Impurities - Gold (Au) ≤ 10.0 ppb
Heavy Metals (as Pb) ≤ 200.0 ppb
Trace Impurities - Iron (Fe) ≤ 10.0 ppb
Trace Impurities - Lead (Pb) ≤ 10.0 ppb
Trace Impurities - Lithium (Li) ≤ 20.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 50.0 ppb
Trace Impurities - Manganese (Mn) ≤ 5.0 ppb
Trace Impurities - Mercury (Hg) ≤ 5.0 ppb
Trace Impurities - Molybdenum (Mo) ≤ 10.0 ppb
Trace Impurities - Nickel (Ni) ≤ 5.0 ppb
Trace Impurities - Niobium (Nb) ≤ 10.0 ppb
Trace Impurities - Potassium (K) ≤ 100 ppb
Trace Impurities - Silicon (Si) ≤ 150.0 ppb
Trace Impurities - Silver (Ag) ≤ 5.0 ppb
Trace Impurities - Sodium (Na) ≤ 100.0 ppb
Trace Impurities - Strontium (Sr) ≤ 10.0 ppb
Trace Impurities - Tantalum (Ta) ≤ 10.0 ppb
Trace Impurities - Thallium (Tl) ≤ 50.0 ppb
Trace Impurities - Tin (Sn) ≤ 20.0 ppb
Trace Impurities - Titanium (Ti) ≤ 10.0 ppb
Trace Impurities - Vanadium (V) ≤ 20.0 ppb
Trace Impurities - Zinc (Zn) ≤ 5.0 ppb
Trace Impurities - Zirconium (Zr) ≤ 10.0 ppb
Particle Count - 0.5 µm and greater ≤ 100 par/ml
Particle Count - 1.0 µm and greater ≤ 10 par/ml
Reported value is the average of all samples counted for this lot number,with no individual sample value exceeding the specification.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR