Anti-PAI-1 (MA-33H1F7) Mouse Monoclonal Antibody [clone: MA-33H1F7]

Lieferant: Merck Millipore (Oncogene)
528217-100
528217-100EA 988 CHF
528217-100
Anti-PAI-1 (MA-33H1F7) Mouse Monoclonal Antibody [clone: MA-33H1F7]
Antikörper
Type: Primary
Antigen: PAI-1 (MA-33H1F7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR