VWR®, Zentrifugenröhrchen, freistehend, mit flachen Schraubverschlüssen
Lieferant: VWR Collection
525-0633EA 211 CHF
525-0633 525-0635 525-0634
VWR®, Zentrifugenröhrchen, freistehend, mit flachen Schraubverschlüssen
Röhrchen Zentrifugenröhrchen
Hergestellt aus hochwertigem, gamma-beständigem, transparentem PP (USP-Klasse 6) mit HDPE-Schraubverschluss.
  • Longer length screw caps
  • Easy-to-read black graduations are accurate to ±2%
  • Large unerasable frosting white printed writing area
  • Both the graduations and writing areas are chloroform-resistant
  • Autoclavable at 121 °C and freezable to –80 °C
  • Leak-Proof
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Diese freistehenden Einmal-Röhrchen verfügen überMore Product Information