Fixed-angle titanium rotor, Type 45 Ti, Optima
Lieferant: Beckman Coulter
339160
521-3712EA 0 CHF
521-3712
Fixed-angle titanium rotor, Type 45 Ti, Optima
Zentrifugen Rotoren
Order Now
TECHNISCHE DATEN
Environmentally Preferable
No. of tubes accepted 6×94 ml / 38×102 mm (6×94 ml / 2×4")
Radius 103,8 mm
Max. RPM 45000