, Sigma-Aldrich®

Lieferant: Merck Millipore (Oncogene)

Synonyms: 4-Nitrophenyl phosphate disodium salt hexahydrate, Phosphatase substrate, p-Nitrophenyl phosphate disodium salt hexahydrate, Phosphoric acid 4-nitrophenyl ester disodium salt hexahydrate, 4-Nitrophenyl phosphate disodium hexahydrate, pNPP disodium salt hexahydrate, di-Sodium 4-nitrophenyl phosphate hexahydrate

4876-1 4876-5
4876-1EA 94.1 CHF
4876-1 4876-5
, Sigma-Aldrich®
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP)
Formel: C₆H₄NNa₂O₆P·6H₂O
Melting Pt: 300 °C
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00066288
CAS-Nummer: 333338-18-4
EINECS: 224-246-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR