Matrix™ Tri-Coded Tubes
Lieferant: THERMO MATRIX TECHNOLOGIES
3741-WP 3741-WP1D 3742-WP1D 3744-WP 3744-WP1D 3745-WP1D 3744-WP-BR 3742-WP1D-BR 3744-WP1D-BR 3741-WP-BR 3741-WP1D-BR 3742-WP-BR 3745-WP1D-BR 3740-WP1D 3744NS-WP1D-BR 3745NS-WP1D-BR 3790-WP1D 3791-WP1D-BR 3792-WP1D-BR
479-1034EA 770 CHF
479-1034 479-1035 479-1039 479-1049 479-1050 479-1055 479-2454 479-2449 479-2450 479-2459 479-2458 479-2460 479-2456 479-2457 479-2453 479-2455 479-2461 479-2462 479-2463
Matrix™ Tri-Coded Tubes
Vials
Matrix™ tri-coded tubes combine the 2D matrix code with an improved linear and human-readable barcode.
  • The linear and human-readable codes match the tubes’ unique 2D code to allow 1D scanning or visual identification of samples across collaborative sites without 2D readers
  • An expanded offering of tri-coded tubes to provide more flexibility for your work
  • The high-contrast barcodes improve the reliability of reading and identifying samples
  • Enhanced sample security with three different barcode formats: 1D (side), human readable (side), 2D barcode (bottom)
More Product Information