Nystatin, Millipore®

Lieferant: Merck Millipore Calbiochem
Achtung

Synonyms: Fungicidin, Mycostatin

475914-1 475914-5
475914-1EA 137 CHF
475914-1 475914-5
Nystatin, Millipore®
Nystatin
Formel: C₄₇H₇₅NO₁₇
Lager Temperatur: Gefrierschrank
MDL Number: MFCD25541651
CAS-Nummer: 1400-61-9
EINECS: 215-749-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR