Eisflockenbereiter, Eigenständig, AF
Lieferant: SCOTSMAN
AF103 AF156 AF124
474-0110EA 4450 CHF
474-0110 474-0112 474-0111
Eisflockenbereiter, Eigenständig, AF
Eisbereiter
The AF self contained ice flakers are air-cooled and manufactured from 304 stainless steel.
  • Integral ice flake storage bin
  • Constructed of high grade stainless steel
  • Ideal for medical and laboratory use
All the models are automatic and produce small irregular flakes of ice with an even melting rate. The flakes are easily moulded to fit into any shape. The ice flakers require a 19 mm 'washing machine' type tapped mains water supply, a correctly rated power supply and a 38 mm vented drain (ideally no higher than the base of the machine), within one metreMore Product Information