Chrom(II)chlorid ≥99,9% (Metall-Basis) -80 mesh

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Chromium dichloride

47176.03 47176.06 47176.14
47176.03EA 105 CHF
47176.03 47176.06 47176.14
Chrom(II)chlorid ≥99,9% (Metall-Basis) -80 mesh
Chrom(II)chlorid

Purity: 99.9% CAS No.: 10049-05-5 Molecular Formula: Cl2Cr Formula Weight: 122.9 Physical Form: -80 Mesh Powder Density: 2.878 Sensitivity: Air and Moisture Sensitive Appearance: Grey Odour: Odourless Melting Point: 824° UN No.: UN3260 MDL No.: MFCD00010947

Formel: Cl₂Cr
Melting Pt: 824 °C
Dichte: 2.878
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00010947
CAS-Nummer: 10049-05-5
EINECS: 233-163-3
UN: 3260
ADR: 8,III
Merck Index: 14,2244

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR