Antimony, plasma standard solution, Sb 10000 µg/ml, Specpure®

Lieferant: Thermo Fisher Scientific

47147.ADEA 169 CHF
47147.AD 47147.AE
Antimony, plasma standard solution, Sb 10000 µg/ml, Specpure®
Standards Kationischer Standards Antimon-Standards

Molecular Formula: Sb in 30% HCl
Physical Form: Liquid
Odour: Acidic
UN No.: UN1789
MDL No.: MFCD00134030

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR