Vakuumtrockenschränke, Vacutherm® Serie
Lieferant: Thermo Scientific
51014539 51014541 51014550 51014542 51014543 51014546 51014547 51014548 51014549 51014552 51014553
466-2710EA 14700 CHF
466-2710 466-2720 466-2700 466-2730 466-2740 466-1107 466-1108 466-1109 466-2741 466-0489 466-0490
Vakuumtrockenschränke, Vacutherm® Serie
Öfen Vakuumtrockenschränke
Vacuum drying ovens with stainless steel chambers, vacuum fittings, tubing and ball valve. Units are designed for gentle drying of heat-sensitive samples and have adjustable temperature limit protection, protection Class 2 (DIN 12880, Part 1).
  • M models, with jacketed heating, have maximum temperature of +200 °C, heat transfer is from vacuum chamber via shelves to the load
  • P models, with shelf heating, have maximum temperature of +300 °C. The shelves incorporate heating coils so heat is transferred directly to the load
  • Rapid heat up times and up to six
More Product Information