Florisil® Pest_Residual, Supelco®

Lieferant: Merck

Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil

MQ100

46382-500G
46382-500GEA 358 CHF
46382-500G
Florisil® Pest_Residual, Supelco®
Magnesiumsilikat
Formel: MgO₃Si
MW: 100.38 g/mol
MDL Number: MFCD00078249
CAS-Nummer: 1343-88-0
EINECS: 215-681-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR