VWR®, External PT100 Probes for
Circulating Baths
Lieferant: VWR Collection
461-1373EA 541 CHF
461-1373 461-1374 461-1376 461-1377
VWR®, External PT100 Probes for
Circulating Baths
Temperaturfühler
External PT100 probes is for monitoring the temperature of an external application.
For use with: Circulating baths, refrigerated circulating baths, heating circulating baths, stainless steel open bath systems, polycarbonate open bath systems, specialty products, calibration bath, 75 L refrigerated circulating bath.