Erbiumtriacetat Tetrahydrat ≥99% (REO, Basis der Oxide der Metalle der seltenen Erden)

Lieferant: Thermo Fisher Scientific

Synonyms: Erbium(III) acetate tetrahydrate

45897.14 45897.22
45897.14EA 170 CHF
45897.14 45897.22
Erbiumtriacetat Tetrahydrat ≥99% (REO, Basis der Oxide der Metalle der seltenen Erden)
Erbiumtriacetat Tetrahydrat

Purity: 99% CAS No.: 15280-57-6 Molecular Formula: C6H9ErO6.4H2O Formula Weight: 416.45 (344.39anhy) Physical Form: Crystalline Aggregates Density: 2.114 Appearance: Pink Odour: Odourless MDL No.: MFCD00013034

Formel: C₆H₉ErO₆·4H₂O
MW: 416.45 g/mol
Dichte: 2.114
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013034
CAS-Nummer: 15280-57-6
EINECS: 247-067-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR