Rhodium ≥99,8% (Metall-Basis), Folie, Dicke 0.025 mm (0.001 in)

Lieferant: Thermo Fisher Scientific

45640.FF
45640.FFEA 0 CHF
45640.FF
Rhodium ≥99,8% (Metall-Basis), Folie, Dicke 0.025 mm (0.001 in)
Rhodium

Purity: 99.8% CAS No.: 7440-16-6 Physical Form: ≈0.2g/25x25mm, certified pinhole free MDL No.: MFCD00011201

Formel: Rh
MW: 102.9 g/mol
Siedepunkt: 3400…3700 °C (1013 hPa)
Melting Pt: 1960
Dichte: 12.41 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011201
CAS-Nummer: 7440-16-6
EINECS: 231-125-0
Merck Index: 12,08353

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR