Eriochromblauschwarz B Indikator für die Komplexometrie (C.I. 14640), Fluka™

Lieferant: Honeywell Chemicals
Achtung

45540-25G
45540-25GEA 73.42 CHF
45540-25G
Eriochromblauschwarz B Indikator für die Komplexometrie (C.I. 14640), Fluka™
Eriochromblauschwarz B
Formel: C₂₀H₁₃N₂NaO₅S
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003934
CAS-Nummer: 3564-14-5
EINECS: 222-639-6

Order Now

Spezifikation Testergebnisse

Appearance of the substance complying
Absorption maximum 523 - 533 nm
Absorbtion (1%/1cm) Min. 100
Suitability for Complexometry complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR