Indium(III)fluorid, anhydrous 99,95 Metals basis

Lieferant: Thermo Fisher Scientific
Achtung

45525.06 45525.14
45525.06EA 210 CHF
45525.06 45525.14
Indium(III)fluorid, anhydrous 99,95 Metals basis
Indium(III)fluorid

Purity: 99.95% CAS No.: 7783-52-0 Molecular Formula: F3In Formula Weight: 171.82 Physical Form: Powder Density: 4.39 Appearance: White Odour: Odourless Melting Point: ca 1170° Boiling Point: 1200° Merck Reference: 14,4959 MDL No.: MFCD00016149

Formel: F₃In
MW: 171.82 g/mol
Siedepunkt: >1200 °C
Melting Pt: ∼ 1170 °C
Dichte: 4.39
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00016149
CAS-Nummer: 7783-52-0
EINECS: 232-005-0
UN: 2923
ADR: 8 II
Merck Index: 14,4959

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR