Graphit ≥99,997% (Metall-Basis), Stäbe, Ø 5.0 mm (0.20 in)

Lieferant: Thermo Fisher Scientific

45076.DW
45076.DWEA 580 CHF
45076.DW
Graphit ≥99,997% (Metall-Basis), Stäbe, Ø 5.0 mm (0.20 in)
Graphit

Purity: 99.997% Physical Form: UF-4S grade MDL No.: MFCD00144065

Formel: C
Melting Pt: 3827 °C
Dichte: 2.09…2.23 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00144065
CAS-Nummer: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR