Dichlormethan, dry_extra 99.9% Stabilised, AcroSeal®

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Methylene chloride

448371000
448371000.EA 63.4 CHF
448371000.
Dichlormethan, dry_extra 99.9% Stabilised, AcroSeal®
Dichlormethan

package of 4x25ML bottles

Formel: CH₂Cl₂
MW: 84.93 g/mol
Siedepunkt: 39…40 °C (760 mmHg)
Melting Pt: –97 °C
Dichte: 1.325 g/cm³
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000881
CAS-Nummer: 75-09-2
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.003 % (Coulometric)
GC ≥99.85 %
Stabilizer Approx. 50 ppm Amylene
Residue after evaporation ≤0.0005 %
Filt.over 0.2 micron PFTE Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR