Nickel 99,5 Metals basis, foil, Dicke 1.0 mm (0.04 in)

Lieferant: Thermo Fisher Scientific
Gefahr

44824.FI 44824.GH 44824.HP
44824.FIEA 83.3 CHF
44824.FI 44824.GH 44824.HP
Nickel 99,5 Metals basis, foil, Dicke 1.0 mm (0.04 in)
Nickel

Purity: 99.5% CAS No.: 7440-02-0 Physical Form: ≈22.3g/50x50mm Odour: Odourless MDL No.: MFCD00011137

Formel: Ni
MW: 58.69 g/mol
Siedepunkt: 3007
Melting Pt: 1453
Dichte: 8.9 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011137
CAS-Nummer: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR