N,N-Dimethylformamid 99.9%, AcroSeal® Biochemistry

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF

447090010 447090025
447090010.EA 86.9 CHF
447090010. 447090025.
N,N-Dimethylformamid 99.9%, AcroSeal® Biochemistry
N,N-Dimethylformamid
Formel: HCON(CH₃)₂
MW: 73.09 g/mol
Siedepunkt: 153 °C (760 mmHg)
Melting Pt: –61 °C
Dichte: 0.945 g/cm³
Flash-Pt: 58 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003284
CAS-Nummer: 68-12-2
UN: 2265
ADR: 3 III
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance Clear liquid
Water ≤0.05 %
GC ≥99.85 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity ≤0.0005 meq/g
Heavy metals (as Pb) ≤5 ppm

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR