2-(Hydroxymethyl)-18-krone-6 ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

44477.01 44477.03
44477.01EA 136 CHF
44477.01 44477.03
2-(Hydroxymethyl)-18-krone-6 ≥97%
2-(Hydroxymethyl)-18-krone-6

Purity: 97% CAS No.: 70069-04-4 Molecular Formula: C13H27O7 Formula Weight: 294.35 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.174 Refractive Index: 1.479 Boiling Point: 150°/0.5mm Beilstein Reference: 4313301 MDL No.: MFCD00188046

Formel: C₁₃H₂₆O₇
Siedepunkt: 150 °C (0.5 mmHg)
Dichte: 1.174
Flash-Pt: >110 °C (230 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00188046
CAS-Nummer: 70069-04-4
EINECS: 274-299-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR