Titan(IV)oxid rutil ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Pigment White 6, Rutile, Titanium dioxide rutile

44375.14 44375.22
44375.14EA 216 CHF
44375.14 44375.22
Titan(IV)oxid rutil ≥99,9% (Metall-Basis)
Titan(IV)oxid rutil

Purity: 99.9% CAS No.: 1317-80-2 Molecular Formula: TiO2 Formula Weight: 79.90 Physical Form: 3-6mm Sintered Pieces Density: 4.23 Melting Point: 1830-1850° Boiling Point: 2500-3000° Merck Reference: 14,9472 MDL No.: MFCD00011269

Formel: O₂Ti
MW: 79.88 g/mol
Siedepunkt: 2500…3000 °C
Melting Pt: 1830…1850 °C
Dichte: 4.23
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011269
CAS-Nummer: 1317-80-2
EINECS: 215-282-2
Merck Index: 14,9472

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR