Tetrabutylammoniumperchlorat elektrochemischer Grad

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Tetra-n-butylammonium perchlorate

43999.09 43999.18 43999.30
43999.09EA 119 CHF
43999.09 43999.18 43999.30
Tetrabutylammoniumperchlorat elektrochemischer Grad
Tetrabutylammonium perchlorate

CAS No.: 1923-70-2 Molecular Formula: C16H36ClNO4 Formula Weight: 341.92 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Melting Point: 210-217° Solubility: Soluble in ethanol. Very slightly soluble in water UN No.: 1481 MDL No.: MFCD00038722


Suitable for use as a polarographic supporting electrolyte

Formel: C₁₆H₃₆ClNO₄
Melting Pt: 210…217 °C
Dichte: 1.07 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00038722
CAS-Nummer: 1923-70-2
EINECS: 217-655-5
UN: 1481
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR