Titan, gauze_wire 18 mesh, Ø 0.28 mm (0.011 in)

Lieferant: Thermo Fisher Scientific
Achtung

43794.GNEA 248 CHF
43794.GN 43794.GY
Titan, gauze_wire 18 mesh, Ø 0.28 mm (0.011 in)
Titanium

CAS No.: 7440-32-6 Physical Form: Wire Cloth Odour: Odourless MDL No.: MFCD00011264

Formel: Ti
MW: 47.88 g/mol
Siedepunkt: 3262
Melting Pt: 1660
Dichte: 4.51 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011264
CAS-Nummer: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR