Trioctylphosphin 90%, AcroSeal®, technical

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: TOP, Tri-n-octylphosphine

436461000
436461000.EA 189 CHF
436461000.
Trioctylphosphin 90%, AcroSeal®, technical
Trioctylphosphin
Formel: C₂₄H₅₁P
MW: 370.64 g/mol
Siedepunkt: 240 °C (10 mmHg)
Melting Pt: 48
Dichte: 0.831 g/cm³
Flash-Pt: 147 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00015298
CAS-Nummer: 4731-53-7
UN: 1760
ADR: 8 II

Order Now

Spezifikation Testergebnisse

Appearance Clear to hazy colorless to yellow Liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
Refractive index 1.4670 to 1.4690 (20°C, 589 nm)
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR