Yttrium(III)-2-ethylhexanoat 10% (w/v) in n-Hexan

Lieferant: Thermo Fisher Scientific
Gefahr

43356.AEEA 267 CHF
43356.AE 43356.AC
Yttrium(III)-2-ethylhexanoat 10% (w/v) in n-Hexan
Yttrium(III)-2-ethylhexanoat

CAS No.: 103470-68-4 Molecular Formula: C24H45O6Y Formula Weight: 518.16 Physical Form: Liquid UN No.: 1208 MDL No.: MFCD00070441

Formel: C₂₄H₄₅O₆Y
MW: 518.524 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00070441
CAS-Nummer: 103470-68-4
UN: 1208
ADR: 3 II

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR