5-Fluoro-2-methylphenylmagnesium bromide 0.5 M in THF, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

431830500
431830500.EA 165 CHF
431830500.
5-Fluoro-2-methylphenylmagnesium bromide 0.5 M in THF, AcroSeal®
5-Fluoro-2-methylphenylmagnesium bromide
Formel: C₇H₆BrFMg
MW: 213.332 g/mol
Dichte: 0.96 g/cm³
Flash-Pt: –17 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01311468
CAS-Nummer: 186496-59-3
UN: 2924
ADR: 3 II

Order Now

Spezifikation Testergebnisse

Appearance Clear yellow Solution
Titration Grignard 0.40 to 0.60 M
Titration with HClO4 ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR