Ammoniumthiocyanat 99,99 Trace metals basis, Supelco®

Lieferant: Merck

Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate

431354-50G
431354-50GEA 0 CHF
431354-50G
Ammoniumthiocyanat 99,99 Trace metals basis, Supelco®
Ammoniumthiocyanat
Formel: NH₄SCN
MW: 76.12 g/mol
Melting Pt: 150
MDL Number: MFCD00011428
CAS-Nummer: 1762-95-4
Merck Index: 13,00564

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR