Lithiumtetrahydroborat 10% (w/w) 4 M in THF, AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Lithium tetrahydridoborate, Lithium borohydride

428888000 428881000
428888000.EA 662 CHF
428888000. 428881000.
Lithiumtetrahydroborat 10% (w/w) 4 M in THF, AcroSeal®
Lithiumtetrahydroborat
Formel: H₄BLi
Siedepunkt: 66 °C (1013 hPa)
Melting Pt: 274 - 277 °C
Dichte: 0.89 g/cm³
Flash-Pt: –21 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011088
CAS-Nummer: 16949-15-8
UN: 3399
ADR: 4.3,I

Order Now

Spezifikation Testergebnisse

Appearance Hazy colorless to grey Solution
Strength 3.8 to 4.3 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR