Kaliumhydroxid standardisierte Lösung 0.1 N in Methanol

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Potash caustic, Caustic potash

42734.K2
42734.K2EA 86.4 CHF
42734.K2
Kaliumhydroxid standardisierte Lösung 0.1 N in Methanol
Kaliumhydroxid

CAS No.: 1310-58-3 Molecular Formula: KOH Formula Weight: 56.11 Physical Form: Liquid Appearance: Colourless Odour: Like alcohol UN No.: 1992 MDL No.: MFCD00003553

Formel: KOH
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003553
CAS-Nummer: 1310-58-3
EINECS: 215-181-3
UN: 1992
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR