Aluminiumoxid, α-Phase ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

Synonyms: ɑ-Tonerde, α-Alumina

42571.22 42571.36 42571.A3
42571.22EA 60.5 CHF
42571.22 42571.36 42571.A3
Aluminiumoxid, α-Phase ≥99,9% (Metall-Basis)
Aluminiumoxid, α-Phase

Purity: 99.9% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 20-50 Micron APS, Spherical Powder, S.A. 5-6m2/g Refractive Index: 1.765 Odour: Odourless Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424

Formel: Al₂O₃
MW: 101.96 g/mol
Siedepunkt: 2980 °C
Melting Pt: 2045 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003424
CAS-Nummer: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR