E/E Untaxed, anhydrous, tested according to Ph.Eur., Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

Synonyms: Ethyl alcohol

41322-1L
41322-1LEA 155.12 CHF
41322-1L
E/E Untaxed, anhydrous, tested according to Ph.Eur., Riedel-de Haën™
Ethanol
Formel: C₂H₅OH
MW: 46.07 g/mol
Siedepunkt: 78…79 °C (1.013 hPa)
Melting Pt: –115 °C
Dichte: 0.790…0.791 g/cm3 (20 °C)
Flash-Pt: 12
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003568
CAS-Nummer: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now

Spezifikation Testergebnisse

Vapor Density 1.59 (vs air)
Appearance complying
Appearance of the solution complying
Assay Min. 99.5 Vol.%
Density (D 20/20) 0.790 - 0.793
Identity (IR) complying
Acidity or alkalinity complying
Non-volatile matter Max. 25 ppm
Residual solvents complying
Volatile impurities (GC) complying
Acetaldehyde & Acetale (GC) Max. 10 ppm
Benzene (GC) Max. 2 ppm
Methanol (GC) Max. 200 ppm
Sum all contaminations (GC) Max. 300 ppm
Absorbance at 240 nm Max. 0.40
Absorbance 250-260 nm Max. 0.30
Absorbance 270-340 nm Max. 0.10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR